4-Chlorophenylurea

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)urea

056984-10G 056984-50G
101911-960EA 120 USD
101911-960 101911-962
4-Chlorophenylurea
4-Chlorophenylurea

Hazard code(s): Irritant

Formula: C₇H₇ClN₂O
MW: 170.6 g/mol
Melting Pt: 204…205 °C
MDL Number: MFCD00014788
CAS Number: 140-38-5
EINECS: 205-412-6

Order Now

Specification Test Results

Melting point 204-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR