1-(4-Chlorophenyl)pyrrole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(4-Chlorophenyl)-1H-pyrrole, N-(4-Chlorophenyl)pyrrole

056980-2G 056980-10G
101911-944EA 150.77 USD
101911-944 101911-946
1-(4-Chlorophenyl)pyrrole ≥98%
1-(4-Chlorophenyl)pyrrole

Hazard code(s): Irritant

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Boiling Pt: 200…203 °C (95 mmHg)
Melting Pt: 86 °C
MDL Number: MFCD00047067
CAS Number: 5044-38-2

Order Now

Specification Test Results

Melting point 86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR