2-Chloro-5-nitrobenzonitrile ≥98%

Supplier: Matrix Scientific

056975-10G 056975-50G
101911-924EA 89.23 USD
101911-924 101911-926
2-Chloro-5-nitrobenzonitrile ≥98%
2-Chloro-5-nitrobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Boiling Pt: 119 - 122 °C (0.6 -rr)
Melting Pt: 108 °C
MDL Number: MFCD00007292
CAS Number: 16588-02-6
EINECS: 240-643-6

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR