2-Chlorobenzamide

Supplier: Matrix Scientific

056944-50G 056944-250G
101911-814EA 116.92 USD
101911-814 101911-816
2-Chlorobenzamide
2-Chlorobenzamide

Hazard code(s): Irritant

Formula: C₇H₆ClNO
Melting Pt: 142…144 °C
Density: 1.34 g/cm³ (18 °C)
MDL Number: MFCD00007973
CAS Number: 609-66-5
EINECS: 210-195-6

Order Now

Specification Test Results

Melting point 142-144°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR