Butyl anthranilate ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: Butyl 2-aminobenzoate, 2-Aminobenzoic acid butyl ester, o-Aminobenzoic acid butyl ester

056936-10G 056936-50G
101911-796EA 183.08 USD
101911-796 101911-798
Butyl anthranilate ≥99%
Butyl anthranilate

Hazard code(s): Irritant

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: ? 0 °C
Density: 1.06 g/cm³ (16 °C)
MDL Number: MFCD00036506
CAS Number: 7756-96-9
EINECS: 231-816-7

Order Now

Specification Test Results

Boiling point 182°C
Density 1.06

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR