1-(4-Bromophenyl)-2-thiourea for synthesis

Supplier: Matrix Scientific
Danger

Synonyms: 1-(4-Bromophenyl)thiourea, N-(4-Bromophenyl)thiourea, 4-Bromophenylthiourea

056931-10G 056931-50G
101911-776EA 261.54 USD
101911-776 101911-778
1-(4-Bromophenyl)-2-thiourea for synthesis
1-(4-Bromophenyl)-2-thiourea

Hazard code(s): Irritant

Formula: (C₆H₄Br)NHC(S)NH₂
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 186 °C
MDL Number: MFCD00041176
CAS Number: 2646-30-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 183-184°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR