3-Bromo-o-toluonitrile ≥98%

Supplier: Matrix Scientific

056922-1G 056922-5G 056922-25G
101911-742EA 315.38 USD
101911-742 101911-744 101911-746
3-Bromo-o-toluonitrile ≥98%
3-Bromo-o-toluonitrile

Hazard code(s): Irritant

Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 44…45 °C
MDL Number: MFCD09025665
CAS Number: 52780-15-1

Order Now

Specification Test Results

Melting point 44-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR