2-Bromo-m-toluonitrile ≥98%

Supplier: Matrix Scientific

056921-1G 056921-5G
101911-738EA 118.46 USD
101911-738 101911-740
2-Bromo-m-toluonitrile ≥98%
2-Bromo-m-toluonitrile

Hazard code(s): Irritant

Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD09025664
CAS Number: 263159-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR