2-Bromo-5-chlorobenzonitrile ≥98%

Supplier: Matrix Scientific

056908-1G 056908-5G
101911-680EA 207.69 USD
101911-680 101911-682
2-Bromo-5-chlorobenzonitrile ≥98%
2-Bromo-5-chlorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃BrClN
MW: 216.46 g/mol
MDL Number: MFCD08445660
CAS Number: 57381-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR