2-Bromo-4-chlorobenzonitrile ≥98%

Supplier: Matrix Scientific

056907-1G 056907-5G 056907-25G
101911-676EA 33.85 USD
101911-676 101911-678 102551-434
2-Bromo-4-chlorobenzonitrile ≥98%
2-Bromo-4-chlorobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₃BrClN
MW: 216.46 g/mol
MDL Number: MFCD00158966
CAS Number: 57381-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR