(4-Biphenyl)acetic acid

Supplier: Matrix Scientific
Danger

Synonyms: 4-Biphenylacetic acid

056896-10G 056896-50G 056896-250G
101911-634EA 89.23 USD
101911-634 101911-636 101911-638
(4-Biphenyl)acetic acid
(4-Biphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₅C₆H₅)CH₂COOH
MW: 212.25 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00004351
CAS Number: 5728-52-9
EINECS: 227-233-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,03979

Order Now

Specification Test Results

Melting point 159-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR