Benzyl-4-hydroxybenzoate

Supplier: Matrix Scientific
Warning

Synonyms: Benzyl 4-hydroxybenzoate, Benzylparaben, 4-Hydroxybenzoic acid benzyl ester, Benzyl Parahydroxybenzoate

056894-25G 056894-100G
101911-624EA 116.92 USD
101911-624 101911-626
Benzyl-4-hydroxybenzoate
Benzyl-4-hydroxybenzoate

Hazard code(s): Irritant

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00016471
CAS Number: 94-18-8
EINECS: 202-311-9

Order Now

Specification Test Results

Melting point 110-112°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR