Anthranilohydrazide

Supplier: Matrix Scientific

Synonyms: 2-Aminobenzoic hydrazide, 2-Aminobenzoic acid hydrazide, 2-Aminobenzohydrazide, 2-Aminobenzoylhydrazide, Anthraniloyl Hydrazine, 2-Aminobenzene-1-carbohydrazide, 2-Aminobenzhydrazide

056885-10G 056885-50G
101911-594EA 69.23 USD
101911-594 101911-596
Anthranilohydrazide
Anthranilohydrazide

Hazard code(s): Irritant

Formula: C₇H₉N₃O
MW: 151.17 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 122…124 °C
MDL Number: MFCD00017056
CAS Number: 1904-58-1
EINECS: 217-604-7

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR