2-Acetyl-1-methyl-1H-pyrrole ≥98%

Supplier: Matrix Scientific

056875-5G 056875-25G 056875-100G
101911-558EA 89.23 USD
101911-558 101911-560 101911-562
2-Acetyl-1-methyl-1H-pyrrole ≥98%
2-Acetyl-1-methyl-1H-pyrrole

Hazard code(s): Irritant

Formula: C₇H₉NO
Boiling Pt: 200 °C (1013 hPa)
Density: 1.0445 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003089
CAS Number: 932-16-1
EINECS: 213-247-6

Order Now

Specification Test Results

Boiling point 200-202°C
Density 1.04


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR