4-Acetoxybenzoic acid

Supplier: Matrix Scientific

056874-25G 056874-100G
101911-554EA 100 USD
101911-554 101911-556
4-Acetoxybenzoic acid
4-Acetoxybenzoic acid

Hazard code(s): Irritant

Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 191…194 °C
MDL Number: MFCD00002540
CAS Number: 2345-34-8
EINECS: 219-066-9

Order Now

Specification Test Results

Melting point 191-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR