2-Octylphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056612-1G 056612-5G
101911-040EA 60 USD
101911-040 101911-042
2-Octylphenyl isothiocyanate ≥98%
2-Octylphenyl isothiocyanate

Hazard code(s): Irritant-Harmful

Formula: C₁₅H₂₁NS
Boiling Pt: 234 °C
Density: 0.90 g/cm³
MDL Number: MFCD00060400
CAS Number: 69626-80-8

Order Now

Specification Test Results

Boiling point 234°C
Density 0.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR