2-Methyl-4-nitrophenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056605-1G 056605-5G
101911-022EA 60 USD
101911-022 101911-024
2-Methyl-4-nitrophenyl isothiocyanate ≥98%
2-Methyl-4-nitrophenyl isothiocyanate

Hazard code(s): Irritant-HARMFUL

Formula: C₈H₆N₂O₂S
MW: 194.21 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD00060542
CAS Number: 135805-96-8

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR