2-Phenylbenzylamine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-(Aminomethyl)biphenyl, [1,1'-Biphenyl]-2-ylmethanamine

056582-1G 056582-5G
101910-950EA 127.69 USD
101910-950 101910-952
2-Phenylbenzylamine ≥98% for synthesis
2-Phenylbenzylamine

Hazard code(s): Irritant

Formula: C₁₃H₁₃N
Boiling Pt: 106 °C (1mm)
Density: 1.06 g/cm³
MDL Number: MFCD00270123
CAS Number: 1924-77-2

Order Now

Specification Test Results

Boiling point 106°C/1mm
Density 1.06


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR