2-Methoxybenzyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056575-1G 056575-5G
101910-928EA 63.08 USD
101910-928 101910-930
2-Methoxybenzyl isothiocyanate ≥98%
2-Methoxybenzyl isothiocyanate

Hazard code(s): Irritant-Harmful, Refrigerate

Formula: C₉H₉NOS
Boiling Pt: 133 °C (4mm)
Melting Pt: 28 °C
MDL Number: MFCD00041111
CAS Number: 17608-09-2

Order Now

Specification Test Results

Boiling point 133°C/4mm
Melting point 28°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR