Cyclohexyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056556-5G 056556-25G
101910-868EA 29.23 USD
101910-868 101910-870
Cyclohexyl isothiocyanate ≥98%
Cyclohexyl isothiocyanate

Hazard code(s): Corrosive, Irritant-Harmful

Formula: C₇H₁₁NS
Boiling Pt: 219 °C (1013 hPa)
Melting Pt: 70 - 72 °C
Density: 1.0339 g/cm³ (20 °C)
MDL Number: MFCD00003841
CAS Number: 1122-82-3
EINECS: 214-361-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 219°C
Density 1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR