3-Bromo-4,5-dihydroxybenzaldehyde ≥95%

Supplier: Matrix Scientific

056469-500MG 056469-1G
101910-620EA 556.92 USD
101910-620 101910-622
3-Bromo-4,5-dihydroxybenzaldehyde ≥95%
3-Bromo-4,5-dihydroxybenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₅BrO₃
MDL Number: MFCD00016608
CAS Number: 16414-34-9
EINECS: 240-463-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR