1-Bromo-3-fluoro-4-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-fluoronitrobenzene

056268-5G 056268-25G
101909-914EA 15.38 USD
101909-914 101909-916
1-Bromo-3-fluoro-4-nitrobenzene ≥95%
1-Bromo-3-fluoro-4-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 52…55 °C
MDL Number: MFCD01930221
CAS Number: 321-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR