2-Bromo-4-methylpyrimidine ≥98%
Supplier: Matrix Scientific
056267-1G
056267-5G
101909-910EA
87.69
USD
101909-910
101909-912
2-Bromo-4-methylpyrimidine ≥98%
2-Bromo-4-methylpyrimidine
Hazard code(s): Irritant
Formula:
C₅H₅ClN₂ |
MDL Number:
MFCD04037883 CAS Number: 130645-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...