2-Chloro-5-nitronicotinic acid ≥95%

Supplier: Matrix Scientific

055440-1G 055440-5G
101909-296EA 315.38 USD
101909-296 101909-298
2-Chloro-5-nitronicotinic acid ≥95%
2-Chloro-5-nitronicotinic acid

Hazard code(s): Irritant

Formula: C₆H₃ClN₂O₄
Melting Pt: 139…140 °C
MDL Number: MFCD06658402
CAS Number: 42959-38-6

Order Now

Specification Test Results

Melting point 139-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR