Piperidin-4-ol ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Piperidinol, 4-Hydroxypiperidine
055401-25G
055401-100G
101909-182EA
55.38
USD
101909-182
101909-184
Piperidin-4-ol ≥98%
Piperidin-4-ol
Hazard code(s): Irritant
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 222 °C (1013 hPa) Melting Pt: 86…89 °C Flash Pt: <100 °C |
MDL Number:
MFCD00005999 CAS Number: 5382-16-1 EINECS: 226-373-1 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...