3,4-Difluorobenzamidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3,4-Difluorophenylamidine hydrochloride, 3,4-Difluorobenzene-1-carboximidamide hydrochloride

054714-500MG 054714-1G
101908-240EA 153.85 USD
101908-240 101908-242
3,4-Difluorobenzamidine hydrochloride ≥95%
3,4-Difluorobenzamidine hydrochloride

Hazard code(s): Irritant

Formula: C₇H₇ClF₂N₂
MW: 192.6 g/mol
MDL Number: MFCD04114434
CAS Number: 107392-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR