3-Bromo-4-chloro-5-nitropyridine ≥97% for synthesis

Supplier: Matrix Scientific

054606-1G 054606-5G
101907-908EA 21.54 USD
101907-908 101907-910
3-Bromo-4-chloro-5-nitropyridine ≥97% for synthesis
3-Bromo-4-chloro-5-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₂BrClN₂O₂
MW: 237.44
Melting Pt: 49…50 °C
MDL Number: MFCD07368836
CAS Number: 31872-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR