3-Bromo-2-chloropyridine-4-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-chloroisonicotinic acid

054389-1G 054389-5G
101907-236EA 110.77 USD
101907-236 101907-238
3-Bromo-2-chloropyridine-4-carboxylic acid ≥98%
3-Bromo-2-chloropyridine-4-carboxylic acid

Hazard code(s): Irritant

Formula: C₆H₃BrClNO₂
MW: 236.449 g/mol
MDL Number: MFCD13185793
CAS Number: 1214323-32-6
EINECS: 217-338-1
UN: 3259
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR