3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98%

Supplier: Matrix Scientific

054384-5G 054384-25G
101907-226EA 27.69 USD
101907-226 101907-228
3-Bromo-α,α,α-trifluoro-5-nitrotoluene ≥98%
3-Bromo-α,α,α-trifluoro-5-nitrotoluene

Hazard code(s): Irritant

Formula: C₇H₃BrF₃NO₂
MW: 270.01 g/mol
MDL Number: MFCD03788719
CAS Number: 630125-49-4

Order Now

Specification Test Results

ND20 1.515

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR