3-Chloro-5-(trifluoromethyl)aniline ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Amino-5-chloro-α,α,α-trifluortoluene, 5-Chloro-α,α,α-trifluoro-m-toluidine, 3-Amino-5-chlorobenzotrifluoride, 3-Amino-5-chloro-α,α,α-trifluorotoluene, 3-Chloro-5-trifluoromethylaniline
054382-1G
054382-5G
054382-25G
101907-216EA
1515.38
USD
101907-216
101907-218
101907-220
3-Chloro-5-(trifluoromethyl)aniline ≥97%
3-Chloro-5-(trifluoromethyl)aniline
Hazard code(s): Irritant
Formula:
C₇H₅ClF₃N MW: 195.57 g/mol |
MDL Number:
MFCD02684210 CAS Number: 69411-05-8 EINECS: 273-989-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...