1-Bromo-2-fluoro-3-nitrobenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-fluoronitrobenzene

054379-1G 054379-5G
101907-202EA 64.62 USD
101907-202 101907-204
1-Bromo-2-fluoro-3-nitrobenzene ≥97%
1-Bromo-2-fluoro-3-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 29…31 °C
MDL Number: MFCD05663754
CAS Number: 58534-94-4

Order Now

Specification Test Results

Melting point 29-31°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR