1-Bromo-3-fluoro-2-nitrobenzene ≥98%

Supplier: Matrix Scientific

054378-1G 054378-5G
101907-198EA 16.92 USD
101907-198 101907-200
1-Bromo-3-fluoro-2-nitrobenzene ≥98%
1-Bromo-3-fluoro-2-nitrobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrFNO₂
Melting Pt: 36…37 °C
MDL Number: MFCD07368788
CAS Number: 886762-70-5

Order Now

Specification Test Results

Melting point 36-37°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR