5-Chloro-2-nitrophenol ≥95%

Supplier: Matrix Scientific

054339-5G 054339-25G
101907-118EA 52.31 USD
101907-118 101907-120
5-Chloro-2-nitrophenol ≥95%
5-Chloro-2-nitrophenol

Hazard code(s): Irritant

Formula: C₆H₄ClNO₃
MDL Number: MFCD09260849
CAS Number: 611-07-4
EINECS: 210-249-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR