8-Bromo-1-octene ≥97%

Supplier: Matrix Scientific
Warning

054187-5G 054187-25G
101906-502EA 53.85 USD
101906-502 101906-504
8-Bromo-1-octene ≥97%
8-Bromo-1-octene

Hazard code(s): Irritant

Formula: C₈H₁₅Br
MW: 191.11 g/mol
Boiling Pt: 198…199 °C
Density: 1.139 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000275
CAS Number: 2695-48-9
EINECS: 220-268-4

Order Now

Specification Test Results

Boiling point 198-199°C
Density 1.139
ND20 1.467

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR