3-Formyl-4-methoxypyridine

Supplier: Matrix Scientific

053825-500MG 053825-2.5G
101905-152EA 230.77 USD
101905-152 101905-154
3-Formyl-4-methoxypyridine
3-Formyl-4-methoxypyridine
Formula: C₇H₇NO₂
MDL Number: MFCD04112505
CAS Number: 82257-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR