4'-Methoxybiphenyl-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 4'-Methoxy[1,1'-biphenyl]-2-carboxylic acid, 4'-Methoxy-[1,1'-biphenyl]-2-carboxylic acid

053748-500MG 053748-2.5G
101904-858EA 364.62 USD
101904-858 101904-860
4'-Methoxybiphenyl-2-carboxylic acid
4'-Methoxybiphenyl-2-carboxylic acid
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD03426469
CAS Number: 18110-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR