2-Chloronicotinamide ≥95%
Supplier: Matrix Scientific
|
Warning
|
052746-5G
052746-25G
101900-872EA
15.38
USD
101900-872
101900-874
2-Chloronicotinamide ≥95%
2-Chloronicotinamide
Hazard code(s): Irritant
Formula:
C₆H₅ClN₂O Melting Pt: 164 - 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006237 CAS Number: 10366-35-5 EINECS: 233-808-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...