2,3-Diaminopyridine ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: Pyridine-2,3-diyldiamine

052745-5G 052745-25G 052745-100G
101900-866EA 29.23 USD
101900-866 101900-868 101900-870
2,3-Diaminopyridine ≥95%
2,3-Diaminopyridine

Hazard code(s): Toxic, Air Sensitive

Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 148…150 °C (5 mmHg)
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00006319
CAS Number: 452-58-4
EINECS: 207-200-9
UN: 2671
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR