4-Amino-3-nitrobenzoic acid ≥97%

Supplier: Matrix Scientific
Warning

052744-25G 052744-100G
101900-862EA 21.54 USD
101900-862 101900-864
4-Amino-3-nitrobenzoic acid ≥97%
4-Amino-3-nitrobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₆N₂O₄
Melting Pt: 290 °C
MDL Number: MFCD00017009
CAS Number: 1588-83-6
EINECS: 216-453-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR