3,5-Dichloroisonicotinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichloropyridine-4-carboxylic acid

048730-5G 048730-25G
101885-036EA 67.69 USD
101885-036 101885-038
3,5-Dichloroisonicotinic acid ≥98%
3,5-Dichloroisonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD01861975
CAS Number: 13958-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR