2-Amino-5-bromo-3-iodopyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-3-iodo-2-pyridylamine, 5-Bromo-3-iodopyridin-2-amine
048725-1G
048725-5G
101885-024EA
15.38
USD
101885-024
101885-026
2-Amino-5-bromo-3-iodopyridine ≥97%
2-Amino-5-bromo-3-iodopyridine
Hazard code(s): Irritant, Light Sensitive
Formula:
C₅H₄BrIN₂ MW: 298.91 g/mol |
MDL Number:
MFCD06659000 CAS Number: 381233-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...