3,4-Dibromopyridine ≥95%

Supplier: Matrix Scientific

048327-5G 048327-25G
101883-736EA 30.77 USD
101883-736 101883-738
3,4-Dibromopyridine ≥95%
3,4-Dibromopyridine

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₃Br₂N
Melting Pt: 71…73 °C
MDL Number: MFCD00234016
CAS Number: 13534-90-2

Order Now

Specification Test Results

Melting point 71-73°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR