3-Azetidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid

048106-5G 048106-25G
101882-822EA 87.69 USD
101882-822 101882-824
3-Azetidinecarboxylic acid ≥95%
3-Azetidinecarboxylic acid

Hazard code(s): Irritant

Formula: C₄H₇NO₂
MW: 101.11 g/mol
Melting Pt: 286…288 °C
MDL Number: MFCD00191763
CAS Number: 36476-78-5

Order Now

Specification Test Results

Melting point 286-288°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR