Methyl-5-bromo-2-methoxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-2-methoxynicotinate, Methyl 5-bromo-2-methoxy-3-pyridinecarboxylate

047945-500MG 047945-1G 047945-5G
101882-098EA 80 USD
101882-098 101882-100 101882-102
Methyl-5-bromo-2-methoxynicotinate ≥95%
Methyl-5-bromo-2-methoxynicotinate

Hazard code(s): Irritant

Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
Melting Pt: 102…104 °C
MDL Number: MFCD08064227
CAS Number: 122433-41-4

Order Now

Specification Test Results

Melting point 102-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR