2,6-Dichloropurine ≥98%

Supplier: Matrix Scientific

047873-25G 047873-100G
101881-790EA 69.23 USD
101881-790 101881-792
2,6-Dichloropurine ≥98%
2,6-Dichloropurine

Hazard code(s): Irritant

Formula: C₅H₂Cl₂N₄
Melting Pt: 184…186 °C
MDL Number: MFCD00077725
CAS Number: 5451-40-1
EINECS: 226-681-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR