3,6-Diiodopyridazine ≥95% for synthesis

Supplier: Matrix Scientific

047829-500MG 047829-1G
101881-618EA 181.54 USD
101881-618 101881-620
3,6-Diiodopyridazine ≥95% for synthesis
3,6-Diiodopyridazine

Hazard code(s): Irritant

Formula: C₄H₂I₂N₂
Melting Pt: 170…172 °C
MDL Number: MFCD01004249
CAS Number: 20698-04-8

Order Now

Specification Test Results

Melting point 170-172°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR