1-(2,4-Dichlorophenyl)-2-thiourea ≥95%
Supplier: Matrix Scientific
047697-500MG
047697-1G
101881-008EA
84.62
USD
101881-008
101881-010
1-(2,4-Dichlorophenyl)-2-thiourea ≥95%
1-(2,4-Dichlorophenyl)-2-thiourea
Hazard code(s): Irritant
Formula:
(C₆H₃Cl₂)NC(S)NH₂ Boiling Pt: 326 °C (1013 hPa) Melting Pt: 160 °C |
MDL Number:
MFCD00022161 CAS Number: 6326-14-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 160°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...