4-Isopropylcinnamic acid ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: 3-(4-Isopropylphenyl)acrylic acid

047689-500MG 047689-1G
101880-988EA 181.54 USD
101880-988 101880-990
4-Isopropylcinnamic acid ≥95%
4-Isopropylcinnamic acid

Hazard code(s): Irritant

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
MDL Number: MFCD00016544
CAS Number: 3368-21-6
EINECS: 222-138-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR