2-Chloro-3,4-dimethoxybenzonitrile ≥95%

Supplier: Matrix Scientific

047672-500MG 047672-1G
101880-932EA 181.54 USD
101880-932 101880-934
2-Chloro-3,4-dimethoxybenzonitrile ≥95%
2-Chloro-3,4-dimethoxybenzonitrile

Hazard code(s): Irritant

Formula: C₉H₈ClNO₂
MDL Number: MFCD00173893
CAS Number: 119413-61-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR