3-Hydroxy-4'-methoxyflavone ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxyflavonol, 3-Hydroxy-2-(4-methoxyphenyl)-4-benzopyrone, 3-Hydroxy-2-(4-methoxyphenyl)chromen-4-one

047600-500MG 047600-1G
101880-732EA 181.54 USD
101880-732 101880-734
3-Hydroxy-4'-methoxyflavone ≥95% for synthesis
3-Hydroxy-4'-methoxyflavone

Hazard code(s): Irritant

Formula: C₁₆H₁₂O₄
MW: 268.27
MDL Number: MFCD00017682
CAS Number: 6889-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR